siRNA Details
| Gene Symbol: Bdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270630.1 | NP_001257559.1 | 2 | 611 | ||
| NM_001270631.1 | NP_001257560.1 | 2 | 762 | |||
| NM_001270632.1 | NP_001257561.1 | 2 | 679 | |||
| NM_001270633.1 | NP_001257562.1 | 2 | 603 | |||
| NM_001270634.1 | NP_001257563.1 | 3 | 490 | |||
| NM_001270635.1 | NP_001257564.1 | 2 | 489 | |||
| NM_001270636.1 | NP_001257565.1 | 2 | 475 | |||
| NM_001270637.1 | NP_001257566.1 | 2 | 467 | |||
| NM_001270638.1 | NP_001257567.1 | 2 | 344 | |||
| NM_012513.4 | NP_036645.2 | 2 | 902 | |||
| XM_006234684.2 | XP_006234746.1 | 2 | 610 | |||
| XM_008762078.1 | XP_008760300.1 | 2 | 2536 | |||
| GenBank mRNA | AY176065.1 | - | 1 | 242 | ||
| AY559248.1 | - | 2 | 405 | |||
| AY559249.1 | - | 2 | 419 | |||
| AY559250.1 | - | 3 | 491 | |||
| BC087634.1 | - | 2 | 597 | 7319966 | ||
| EF125675.1 | - | 2 | 902 | |||
| EF125676.1 | - | 2 | 468 | |||
| EF125677.1 | - | 2 | 680 | |||
| EF125678.1 | - | 2 | 763 | |||
| EF125679.1 | - | 2 | 603 | |||
| EF125680.1 | - | 2 | 611 | |||
| EF125686.1 | - | 2 | 489 | |||
| EF125687.1 | - | 2 | 344 | |||
| EF125688.1 | - | 2 | 476 | |||
| EF125689.1 | - | 2 | 540 | |||
| EF125690.1 | - | 1 | 734 | |||
| M61175.1 | - | 2 | 314 | |||
| X67108.1 | - | 1 | 263 | |||

Bdnf
Genomic Map