siRNA Details
| Gene Symbol: Cpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012533.1 | NP_036665.1 | 2 | 128 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpb1| Gene Symbol: Cpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012533.1 | NP_036665.1 | 2 | 128 | ||