siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012557.2 | NP_036689.2 | 2 | 392 | ||
| XM_006253488.2 | XP_006253550.1 | 3 | 667 | |||
| XM_006253489.2 | XP_006253551.1 | 2 | 524 | |||
| XM_006253490.2 | XP_006253552.1 | 2 | 369 | |||
| GenBank mRNA | BC168147.1 | - | 2 | 392 | 9024689 | |
| U73586.1 | - | 1 | 168 | |||

Fancc
Genomic Map