siRNA Details
| Gene Symbol: Gja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012567.2 | NP_036699.1 | 2 | 506 | ||
| XM_006256503.2 | XP_006256565.1 | 2 | 389 | |||
| GenBank mRNA | AY324140.1 | - | 2 | 506 | ||
| BC081842.1 | - | 2 | 502 | 7105924 | ||

Gja1
Genomic Map