siRNA Details
| Gene Symbol: Add2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109880.1 | NP_001103350.1 | 4 | 400 | ||
| NM_012491.2 | NP_036623.1 | 4 | 400 | |||
| GenBank mRNA | M63894.1 | - | 4 | 400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Add2| Gene Symbol: Add2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109880.1 | NP_001103350.1 | 4 | 400 | ||
| NM_012491.2 | NP_036623.1 | 4 | 400 | |||
| GenBank mRNA | M63894.1 | - | 4 | 400 | ||