siRNA Details
| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012500.1 | NP_036632.1 | 4 | 361 | ||
| XM_008766476.1 | XP_008764698.1 | 3 | 465 | |||
| GenBank mRNA | BC166605.1 | - | 4 | 367 | 7937766 | |
| J04733.1 | - | 4 | 361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apeh| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012500.1 | NP_036632.1 | 4 | 361 | ||
| XM_008766476.1 | XP_008764698.1 | 3 | 465 | |||
| GenBank mRNA | BC166605.1 | - | 4 | 367 | 7937766 | |
| J04733.1 | - | 4 | 361 | |||