siRNA Details
| Gene Symbol: Cacna1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012517.2 | NP_036649.2 | 1 | 270 | ||
| XM_006237159.2 | XP_006237221.1 | 1 | 94 | |||
| XM_006237170.2 | XP_006237232.1 | 1 | 115 | |||
| XM_006237171.2 | XP_006237233.1 | 1 | 115 | |||
| XM_006237172.2 | XP_006237234.1 | 1 | 115 | |||
| GenBank mRNA | AF394938.1 | - | 1 | 121 | ||
| AF394939.1 | - | 1 | 121 | |||
| AF394940.1 | - | 1 | 121 | |||
| M59786.1 | - | 1 | 273 | |||

Cacna1c
Genomic Map