siRNA Details
| Gene Symbol: Ace | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012544.1 | NP_036676.1 | 2 | 355 | ||
| GenBank mRNA | AF201331.1 | - | 2 | 355 | ||
| AF201332.1 | - | 2 | 355 | |||
| AF532783.1 | - | 2 | 299 | |||
| AF532784.1 | - | 2 | 299 | |||
| BC085760.1 | - | 2 | 355 | 7097136 | ||
| U03708.1 | - | 2 | 355 | |||
| U03734.1 | - | 2 | 355 | |||

Ace
Genomic Map