siRNA Details
| Gene Symbol: Ednra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012550.2 | NP_036682.1 | 5 | 1065 | ||
| XM_006255420.2 | XP_006255482.1 | 5 | 1231 | |||
| XM_008772471.1 | XP_008770693.1 | 5 | 938 | |||
| XM_008772472.1 | XP_008770694.1 | 4 | 1133 | |||
| GenBank mRNA | DQ832324.1 | - | 4 | 816 | ||
| DQ839232.1 | - | 3 | 671 | |||
| DQ839233.1 | - | 5 | 873 | |||
| DQ839234.1 | - | 5 | 931 | |||
| M60786.1 | - | 4 | 816 | |||

Ednra
Genomic Map