siRNA Details
| Gene Symbol: Foxg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012560.2 | NP_036692.1 | 1 | 1175 | ||
| GenBank mRNA | M87634.1 | - | 1 | 1176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxg1| Gene Symbol: Foxg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012560.2 | NP_036692.1 | 1 | 1175 | ||
| GenBank mRNA | M87634.1 | - | 1 | 1176 | ||