siRNA Details
| Gene Symbol: Fuca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012562.2 | NP_036694.2 | 2, 3 | 541 | ||
| GenBank mRNA | BC081844.1 | - | 2, 3 | 541 | 7192716 | |
| X16145.1 | - | 2, 3 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fuca1| Gene Symbol: Fuca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012562.2 | NP_036694.2 | 2, 3 | 541 | ||
| GenBank mRNA | BC081844.1 | - | 2, 3 | 541 | 7192716 | |
| X16145.1 | - | 2, 3 | 531 | |||