siRNA Details
| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012574.1 | NP_036706.1 | 3 | 898 | ||
| GenBank mRNA | M91562.1 | - | 3 | 605 | ||
| U11419.1 | - | 3 | 898 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin2b| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012574.1 | NP_036706.1 | 3 | 898 | ||
| GenBank mRNA | M91562.1 | - | 3 | 605 | ||
| U11419.1 | - | 3 | 898 | |||