siRNA Details
| Gene Symbol: Apc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012499.1 | NP_036631.1 | 15 | 6620 | ||
| XM_006254506.2 | XP_006254568.1 | 16 | 6880 | |||
| XM_006254508.2 | XP_006254570.1 | 17 | 6721 | |||
| XM_006254509.2 | XP_006254571.1 | 17 | 6690 | |||
| XM_006254510.2 | XP_006254572.1 | 15 | 6826 | |||
| XM_006254511.2 | XP_006254573.1 | 16 | 6667 | |||
| XM_006254512.2 | XP_006254574.1 | 16 | 7329 | |||
| XM_006254513.2 | XP_006254575.1 | 15 | 6775 | |||
| XM_006254515.2 | XP_006254577.1 | 17 | 6546 | |||
| XM_006254518.2 | XP_006254580.1 | 16 | 7144 | |||
| XM_006254520.2 | XP_006254582.1 | 16 | 6577 | |||
| XM_006254521.2 | XP_006254583.1 | 16 | 6364 | |||
| XM_008772015.1 | XP_008770237.1 | 16 | 8812 | |||
| XM_008772016.1 | XP_008770238.1 | 16 | 8762 | |||
| XM_008772017.1 | XP_008770239.1 | 16 | 7153 | |||
| GenBank mRNA | D38629.1 | - | 15 | 6620 | ||

Apc
Genomic Map