siRNA Details
| Gene Symbol: Asgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012503.2 | NP_036635.1 | 8, 9 | 764 | ||
| XM_006246579.1 | XP_006246641.1 | 8, 9 | 876 | |||
| XM_008767768.1 | XP_008765990.1 | 8, 9 | 896 | |||
| XM_008767769.1 | XP_008765991.1 | 8, 9 | 840 | |||
| GenBank mRNA | BC088154.1 | - | 8, 9 | 764 | 7372227 | |
| FQ218553.1 | - | 8, 9 | 840 | |||
| FQ218644.1 | - | 8, 9 | 800 | |||
| FQ218824.1 | - | 8, 9 | 800 | |||
| FQ219710.1 | - | 8, 9 | 881 | |||
| M21770.1 | - | 7, 8 | 666 | |||

Asgr1
Genomic Map