siRNA Details
| Gene Symbol: Atp7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012511.2 | NP_036643.2 | 2 | 835 | ||
| XM_006253346.2 | XP_006253408.1 | 2 | 1511 | |||
| GenBank mRNA | AB017790.1 | - | 2 | 835 | ||
| AB017791.1 | - | 2 | 835 | |||
| AB017792.1 | - | 2 | 835 | |||
| AB017793.1 | - | 2 | 835 | |||
| U08344.1 | - | 2 | 851 | |||

Atp7b
Genomic Map