siRNA Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012529.3 | NP_036661.3 | 832 | |||
| GenBank mRNA | BC065307.1 | - | 814 | 5598839 | ||
| BC070955.1 | - | 819 | 7104205 | |||
| BC087656.1 | - | 831 | ||||
| BC099814.1 | - | 816 | 7929525 | |||
| BC127477.1 | - | 822 | 8367180 | |||
| FQ213834.1 | - | 576 | ||||
| FQ216091.1 | - | 4 | 551 | |||
| M14400.1 | - | 844 | ||||
| M57664.1 | - | 5 | 695 | |||

Ckb
Genomic Map