siRNA Details
| Gene Symbol: Ddc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270852.1 | NP_001257781.1 | 10, 11 | 1062 | ||
| NM_001270853.1 | NP_001257782.1 | 9, 10 | 956 | |||
| NM_012545.4 | NP_036677.1 | 10, 11 | 1102 | |||
| XM_006251474.2 | XP_006251536.1 | 10, 11 | 1838 | |||
| XM_006251475.2 | XP_006251537.1 | 10, 11 | 1678 | |||
| XM_008770251.1 | XP_008768473.1 | 10, 11 | 1133 | |||
| XM_008770252.1 | XP_008768474.1 | 10, 11 | 1196 | |||
| XM_008770253.1 | XP_008768475.1 | 9, 10 | 1013 | |||
| XM_008770254.1 | XP_008768476.1 | 10, 11 | 1133 | |||
| GenBank mRNA | BC087032.1 | - | 10, 11 | 1062 | 7098083 | |
| FQ219697.1 | - | 10, 11 | 1100 | |||
| JF273828.1 | - | 9, 10 | 956 | |||
| M27716.1 | - | 10, 11 | 1107 | |||
| U31884.1 | - | 10, 11 | 1084 | |||

Ddc
Genomic Map