siRNA Details
| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012570.2 | NP_036702.1 | 10 | 1478 | ||
| GenBank mRNA | BC081841.1 | - | 10 | 1362 | 7107138 | |
| FQ214067.1 | - | 2 | 58 | |||
| U95148.1 | - | 1 | 19 | |||
| X14044.1 | - | 10 | 1368 | |||
| X14223.1 | - | 10 | 1341 | |||

Glud1
Genomic Map