siRNA Details
| Gene Symbol: H1f0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012578.2 | NP_036710.1 | 1 | 656 | ||
| GenBank mRNA | BC061842.1 | - | 1 | 656 | 5621199 | |
| X70685.1 | - | 1 | 426 | |||
| X72624.1 | - | 1 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H1f0| Gene Symbol: H1f0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012578.2 | NP_036710.1 | 1 | 656 | ||
| GenBank mRNA | BC061842.1 | - | 1 | 656 | 5621199 | |
| X70685.1 | - | 1 | 426 | |||
| X72624.1 | - | 1 | 534 | |||