siRNA Details
| Gene Symbol: Irf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012591.1 | NP_036723.1 | 8 | 970 | ||
| GenBank mRNA | BC076382.1 | - | 8 | 883 | 7099391 | |
| M34253.1 | - | 8 | 970 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irf1| Gene Symbol: Irf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012591.1 | NP_036723.1 | 8 | 970 | ||
| GenBank mRNA | BC076382.1 | - | 8 | 883 | 7099391 | |
| M34253.1 | - | 8 | 970 | |||