siRNA Details
| Gene Symbol: Lepr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012596.1 | NP_036728.1 | 3 | 368 | ||
| GenBank mRNA | AF287268.1 | - | 2 | 294 | ||
| D84125.1 | - | 2 | 294 | |||
| D84126.1 | - | 2 | 294 | |||
| D84550.1 | - | 2 | 294 | |||
| D84551.1 | - | 2 | 294 | |||
| D85558.1 | - | 2 | 314 | |||
| U52966.1 | - | 3 | 368 | |||
| U53144.1 | - | 3 | 411 | |||
| U60151.1 | - | 2 | 294 | |||

Lepr
Genomic Map