siRNA Details
| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012600.2 | NP_036732.2 | 1, 2 | 86 | ||
| GenBank mRNA | FQ228526.1 | - | 1, 2 | 117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Me1| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012600.2 | NP_036732.2 | 1, 2 | 86 | ||
| GenBank mRNA | FQ228526.1 | - | 1, 2 | 117 | ||