siRNA Details
| Gene Symbol: Nf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012609.1 | NP_036741.1 | 218 | |||
| GenBank mRNA | D45201.1 | - | 218 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010897, AK085050, BB660074, L10370.

Nf1
Genomic Map