siRNA Details
| Gene Symbol: Odc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239906.2 | 4 | 484 | |||
| XM_006239907.2 | XP_006239969.1 | 4 | 474 | |||
| XM_006239908.2 | XP_006239970.1 | 3 | 390 | |||
| XM_008764573.1 | 4 | 457 | ||||
| GenBank mRNA | BC078882.1 | - | 2 | 162 | 7115603 | |
| CB613128.1 | - | 4 | 541 | |||
| J04791.1 | - | 4 | 278 | |||
| M16982.1 | - | 4 | 542 | |||

Odc1
Genomic Map