siRNA Details
| Gene Symbol: Sdc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012649.2 | NP_036781.1 | 2 | 141 | ||
| GenBank mRNA | M81786.1 | - | 2 | 141 | ||
| S61868.1 | - | 2 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdc4| Gene Symbol: Sdc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012649.2 | NP_036781.1 | 2 | 141 | ||
| GenBank mRNA | M81786.1 | - | 2 | 141 | ||
| S61868.1 | - | 2 | 141 | |||