siRNA Details
| Gene Symbol: Abcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012690.2 | NP_036822.1 | 4 | 185 | ||
| XM_008762697.1 | XP_008760919.1 | 4 | 367 | |||
| GenBank mRNA | L15079.1 | - | 4 | 187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb4| Gene Symbol: Abcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012690.2 | NP_036822.1 | 4 | 185 | ||
| XM_008762697.1 | XP_008760919.1 | 4 | 367 | |||
| GenBank mRNA | L15079.1 | - | 4 | 187 | ||