siRNA Details
| Gene Symbol: Lpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012598.2 | NP_036730.1 | 3 | 453 | ||
| GenBank mRNA | BC081836.1 | - | 3 | 453 | 7106121 | |
| L03294.1 | - | 438 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpl| Gene Symbol: Lpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012598.2 | NP_036730.1 | 3 | 453 | ||
| GenBank mRNA | BC081836.1 | - | 3 | 453 | 7106121 | |
| L03294.1 | - | 438 | ||||