siRNA Details
| Gene Symbol: Nf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012609.1 | NP_036741.1 | 262 | |||
| GenBank mRNA | D45201.1 | - | 262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nf1| Gene Symbol: Nf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012609.1 | NP_036741.1 | 262 | |||
| GenBank mRNA | D45201.1 | - | 262 | |||