siRNA Details
| Gene Symbol: Omp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012616.1 | NP_036748.1 | 1 | 478 | ||
| GenBank mRNA | M15644.1 | - | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Omp| Gene Symbol: Omp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012616.1 | NP_036748.1 | 1 | 478 | ||
| GenBank mRNA | M15644.1 | - | 416 | |||