siRNA Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012619.2 | NP_036751.2 | 2, 3 | 265 | ||
| GenBank mRNA | BC078881.1 | - | 2, 3 | 265 | 7131414 | |
| FQ219680.1 | - | 2, 3 | 295 | |||
| M12337.1 | - | 2, 3 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pah| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012619.2 | NP_036751.2 | 2, 3 | 265 | ||
| GenBank mRNA | BC078881.1 | - | 2, 3 | 265 | 7131414 | |
| FQ219680.1 | - | 2, 3 | 295 | |||
| M12337.1 | - | 2, 3 | 250 | |||