siRNA Details
| Gene Symbol: Pklr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012624.3 | NP_036756.3 | 3 | 261 | ||
| XM_006232593.2 | XP_006232655.1 | 3 | 392 | |||
| GenBank mRNA | BC161827.1 | - | 2 | 251 | 7127842 | |
| M11709.1 | - | 3 | 262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pklr| Gene Symbol: Pklr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012624.3 | NP_036756.3 | 3 | 261 | ||
| XM_006232593.2 | XP_006232655.1 | 3 | 392 | |||
| GenBank mRNA | BC161827.1 | - | 2 | 251 | 7127842 | |
| M11709.1 | - | 3 | 262 | |||