siRNA Details
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012628.1 | NP_036760.1 | 4, 5 | 607 | ||
XM_006228014.2 | XP_006228076.1 | 4, 5 | 675 | |||
GenBank mRNA | BC089226.1 | - | 4, 5 | 641 | 7319243 | |
M13707.1 | - | 4, 5 | 607 | |||
X07287.1 | - | 4, 5 | 530 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
