siRNA Details
| Gene Symbol: Prl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012629.1 | NP_036761.1 | 4 | 632 | ||
| XM_006253912.2 | XP_006253974.1 | 4 | 524 | |||
| GenBank mRNA | AF022935.1 | - | 4 | 632 | ||
| BC166892.1 | - | 4 | 497 | 6919622 | ||
| BC168729.1 | - | 4 | 503 | 6922061 | ||
| S46794.1 | - | 3 | 368 | |||
| V01249.1 | - | 4 | 497 | |||
| V01250.1 | - | 3 | 412 | |||

Prl
Genomic Map