siRNA Details
| Gene Symbol: Shbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012650.1 | NP_036782.1 | 6 | 855 | ||
| XM_006246586.2 | XP_006246648.1 | 7 | 1604 | |||
| XM_006246592.2 | XP_006246654.1 | 6 | 962 | |||
| XM_008767771.1 | XP_008765993.1 | 7 | 1069 | |||
| XM_008767772.1 | XP_008765994.1 | 6 | 877 | |||
| GenBank mRNA | BC097336.1 | - | 6 | 843 | 7453185 | |
| M15034.1 | - | 6 | 855 | |||
| M31179.1 | - | 6 | 807 | |||

Shbg
Genomic Map