siRNA Details
| Gene Symbol: Tgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012671.2 | NP_036803.1 | 3 | 344 | ||
| XM_006236822.2 | XP_006236884.1 | 3 | 1237 | |||
| XM_008763049.1 | XP_008761271.1 | 3 | 1241 | |||
| XM_008763050.1 | XP_008761272.1 | 3 | 1238 | |||
| GenBank mRNA | KF366251.1 | - | 3 | 200 | ||
| M31076.1 | - | 3 | 344 | |||
| X02004.1 | - | 4 | 344 | |||
| X61485.1 | - | 2 | 125 | |||
| X61487.1 | - | 2 | 125 | |||

Tgfa
Genomic Map