siRNA Details
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270854.1 | NP_001257783.1 | 6 | 570 | ||
NM_012672.3 | NP_036804.2 | 6 | 570 | |||
XM_006251709.2 | XP_006251771.1 | 2 | 591 | |||
XM_008770500.1 | XP_008768722.1 | 4 | 482 | |||
GenBank mRNA | DQ191165.1 | - | 3 | 317 | ||
HM043807.1 | - | 2 | 359 | |||
J03819.1 | - | 6 | 566 | |||
J03933.1 | - | 6 | 567 |
