siRNA Details
| Gene Symbol: Ldhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012595.1 | NP_036727.1 | 6 | 720 | ||
| XM_006237615.2 | XP_006237677.1 | 7 | 891 | |||
| GenBank mRNA | BC059149.1 | - | 6 | 777 | 6919339 | |
| FQ212899.1 | - | 6 | 783 | |||
| FQ214847.1 | - | 6 | 767 | |||
| FQ215687.1 | - | 6 | 783 | |||
| U07181.1 | - | 6 | 720 | |||

Ldhb
Genomic Map