siRNA Details
| Gene Symbol: Lepr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012596.1 | NP_036728.1 | 19 | 3181 | ||
| GenBank mRNA | AF287268.1 | - | 18 | 3107 | ||
| D84550.1 | - | 18 | 3107 | |||
| D84551.1 | - | 18 | 3107 | |||
| D85558.1 | - | 18 | 3127 | |||
| U52966.1 | - | 19 | 3181 | |||
| U60151.1 | - | 18 | 3107 | |||

Lepr
Genomic Map