siRNA Details
| Gene Symbol: S100a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012618.2 | NP_036750.1 | 3 | 257 | ||
| XM_006232592.2 | XP_006232654.1 | 3 | 317 | |||
| GenBank mRNA | FQ215053.1 | - | 3 | 257 | ||
| FQ220722.1 | - | 3 | 257 | |||
| FQ221062.1 | - | 3 | 257 | |||
| FQ221355.1 | - | 3 | 257 | |||
| FQ221406.1 | - | 3 | 258 | |||
| FQ221428.1 | - | 3 | 257 | |||
| FQ221508.1 | - | 3 | 257 | |||
| FQ221699.1 | - | 3 | 260 | |||
| FQ221706.1 | - | 3 | 258 | |||
| FQ221757.1 | - | 3 | 258 | |||
| FQ221860.1 | - | 3 | 258 | |||
| FQ222027.1 | - | 3 | 258 | |||
| FQ222069.1 | - | 3 | 257 | |||
| FQ222186.1 | - | 3 | 257 | |||
| FQ222219.1 | - | 3 | 258 | |||
| FQ222585.1 | - | 3 | 257 | |||
| FQ222875.1 | - | 3 | 257 | |||
| FQ222962.1 | - | 3 | 258 | |||
| FQ223009.1 | - | 3 | 257 | |||
| FQ223047.1 | - | 3 | 261 | |||
| FQ223108.1 | - | 3 | 240 | |||
| FQ223128.1 | - | 3 | 258 | |||
| FQ223234.1 | - | 3 | 257 | |||
| FQ228364.1 | - | 3 | 259 | |||
| FQ228506.1 | - | 3 | 257 | |||
| FQ228607.1 | - | 3 | 259 | |||
| FQ229007.1 | - | 3 | 256 | |||
| FQ229874.1 | - | 3 | 257 | |||
| J03628.1 | - | 3 | 258 | |||
| X64022.1 | - | 2 | 200 | |||
| X64023.1 | - | 13 | ||||

S100a4
Genomic Map