siRNA Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012639.2 | NP_036771.1 | 12, 13 | 1666 | ||
| XM_008763217.1 | XP_008761439.1 | 13, 14 | 1796 | |||
| XM_008763218.1 | XP_008761440.1 | 13, 14 | 1976 | |||
| GenBank mRNA | BC062071.1 | - | 12, 13 | 1666 | 5599540 | |
| M15427.1 | - | 1410 | ||||

Raf1
Genomic Map