siRNA Details
| Gene Symbol: Thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270854.1 | NP_001257783.1 | 6, 7 | 645 | ||
| XM_006251709.2 | XP_006251771.1 | 2, 3 | 666 | |||
| XM_008770500.1 | XP_008768722.1 | 4, 5 | 557 | |||
| GenBank mRNA | J03819.1 | - | 6, 7 | 641 | ||
| J03933.1 | - | 6, 7 | 642 | |||

Thrb
Genomic Map