siRNA Details
| Gene Symbol: Gipr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012714.1 | NP_036846.1 | 5, 6 | 549 | ||
| GenBank mRNA | L19660.1 | - | 5, 6 | 549 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gipr| Gene Symbol: Gipr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012714.1 | NP_036846.1 | 5, 6 | 549 | ||
| GenBank mRNA | L19660.1 | - | 5, 6 | 549 | ||
Set Membership: |
|