siRNA Details
| Gene Symbol: P2rx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012721.2 | NP_036853.2 | 2 | 303 | ||
| XM_008768851.1 | XP_008767073.1 | 2 | 493 | |||
| XM_008768852.1 | XP_008767074.1 | 2 | 178 | |||
| GenBank mRNA | X92070.1 | - | 2 | 184 | ||
| X97376.1 | - | 2 | 314 | |||

P2rx6
Genomic Map