siRNA Details
| Gene Symbol: Klkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012725.2 | NP_036857.2 | 3, 4 | 279 | ||
| XM_006253121.2 | XP_006253183.1 | 3, 4 | 1213 | |||
| XM_008771252.1 | XP_008769474.1 | 1, 2 | 34 | |||
| XM_008771253.1 | XP_008769475.1 | 1, 2 | 128 | |||
| GenBank mRNA | BC089815.1 | - | 3, 4 | 279 | 7369524 | |
| M30282.1 | - | 3, 4 | 242 | |||
| M58590.1 | - | 3, 4 | 242 | |||

Klkb1
Genomic Map