siRNA Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012727.3 | NP_036859.2 | 6 | 591 | ||
| XM_006254523.2 | XP_006254585.1 | 5 | 533 | |||
| GenBank mRNA | BC128706.1 | - | 6 | 591 | 7455982 | |
| M63334.1 | - | 485 | ||||
| M64757.1 | - | 4 | 581 | |||
| S65840.1 | - | 5 | 516 | |||

Camk4
Genomic Map