siRNA Details
| Gene Symbol: Hk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012735.2 | NP_036867.1 | 2 | 339 | ||
| GenBank mRNA | AF027179.1 | - | 2 | 143 | ||
| M68971.1 | - | 2 | 340 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hk2| Gene Symbol: Hk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012735.2 | NP_036867.1 | 2 | 339 | ||
| GenBank mRNA | AF027179.1 | - | 2 | 143 | ||
| M68971.1 | - | 2 | 340 | |||