siRNA Details
| Gene Symbol: Gpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012736.1 | NP_036868.1 | 2, 3 | 194 | ||
| GenBank mRNA | BC083565.1 | - | 2, 3 | 152 | 7134834 | |
| FQ223746.1 | - | 450 | ||||
| U08027.1 | - | 2, 3 | 173 | |||
| X78593.1 | - | 2, 3 | 194 | |||

Gpd2
Genomic Map