siRNA Details
| Gene Symbol: Fyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012755.1 | NP_036887.1 | 4, 5 | 479 | ||
| XM_006256520.2 | XP_006256582.1 | 2, 3 | 1314 | |||
| XM_006256521.2 | XP_006256583.1 | 2, 3 | 1314 | |||
| XM_006256522.1 | XP_006256584.1 | 3, 4 | 766 | |||
| GenBank mRNA | U35365.1 | - | 4, 5 | 479 | ||

Fyn
Genomic Map