siRNA Details
| Gene Symbol: Mas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012757.2 | NP_036889.1 | 4 | 704 | ||
| XM_006227859.2 | XP_006227921.1 | 3 | 1966 | |||
| XM_006227860.2 | XP_006227922.1 | 3 | 2088 | |||
| XM_006227867.2 | XP_006227929.1 | 2 | 1777 | |||
| XM_006227868.2 | XP_006227930.1 | 2 | 1725 | |||
| XM_008758726.1 | XP_008756948.1 | 3 | 2085 | |||
| XM_008758727.1 | XP_008756949.1 | 3 | 2036 | |||
| XM_008758728.1 | XP_008756950.1 | 3 | 1732 | |||
| XM_008758729.1 | XP_008756951.1 | 5 | 1021 | |||
| XM_008758730.1 | XP_008756952.1 | 3 | 2033 | |||
| GenBank mRNA | BC078884.1 | - | 4 | 704 | 7131760 | |

Mas1
Genomic Map