siRNA Details
| Gene Symbol: Gpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012774.1 | NP_036906.1 | 1 | 230 | ||
| GenBank mRNA | BC085756.1 | - | 1 | 245 | 7122343 | |
| M22400.1 | - | 1 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc3| Gene Symbol: Gpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012774.1 | NP_036906.1 | 1 | 230 | ||
| GenBank mRNA | BC085756.1 | - | 1 | 245 | 7122343 | |
| M22400.1 | - | 1 | 230 | |||